BrettPhos palladacycle 3rd generation 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulphonate, BrettPhos Pd G3

047250-03 047250-06 047250-MD
AA47250-03EA 478 USD
AA47250-03 AA47250-06 AA47250-MD
BrettPhos palladacycle 3rd generation 98%
BrettPhos palladacycle 3rd generation

Chloro(2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl)(2'-aminobiphenyl-2-yl)palladium(II) methanesulfonate, 98%


Purity: 98% CAS No.: 1470372-59-8 Molecular Formula: C48H66NO5PPdS Formula Weight: 906.50 Physical Form: Solid Appearance: Yellow MDL No.: MFCD22572666

Formula: C₄₈H₆₆NO₅PPdS
MW: 906.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22572666
CAS Number: 1470372-59-8

Order Now

Specification Test Results

Appearance (Color) Yellow
Form Powder
Elemental Analysis 62.0 to 65.0 wt% Carbon

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR