Iron(III) hydroxide, α-phase 20% in water stabilised, nanoparticle dispersion, Diameter <5.0 nm APS Number Weighted

Supplier: Thermo Scientific Chemicals
Warning

047241-22 047241-36
AA47241-22EA 193 USD
AA47241-22 AA47241-36
Iron(III) hydroxide, α-phase 20% in water stabilised, nanoparticle dispersion, Diameter <5.0 nm APS Number Weighted
Iron(III) hydroxide, alpha-phase

Iron(III) hydroxide, 20% in H{2}O, nanoparticle dispersion, low pH, 5.0nm APS Number Weighted


CAS No.: 20344-49-4 Molecular Formula: FeHO2 Formula Weight: 88.85 Physical Form: Liquid Density: 1.20 Appearance: Red-brown Odor: Odorless pH: 3.0 MDL No.: MFCD00064782

Formula: FeO(OH)
MW: 88.85 g/mol
Density: 1.20
Storage Temperature: Ambient
MDL Number: MFCD00064782
CAS Number: 20344-49-4
EINECS: 243-746-4
Merck Index: 14,4025

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR