Copper bis(trifluoromethylsulfonyl)imide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper bis(trifluoromethanesulphonimide), Copper bis(trifluoromethanesulphonyl)imide

047162-03 047162-06 047162-MD
AA47162-03EA 134 USD
AA47162-03 AA47162-06 AA47162-MD
Copper bis(trifluoromethylsulfonyl)imide
Copper bis(trifluoromethylsulfonyl)imide

Copper bis(trifluoromethylsulfonyl)imide


CAS No.: 291300-50-0 Molecular Formula: C2CuF6NO4S2 Formula Weight: 343.68 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Blue UN No.: 3261 MDL No.: MFCD23380170

Formula: C₂CuF₆NO₄S₂
MW: 343.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380170
CAS Number: 291300-50-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR