Tin bis(trifluoromethylsulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tin bis(trifluoromethanesulphonimide), Tin bis(trifluoromethanesulphonyl)imide
047142-03
047142-06
047142-MD
AA47142-03EA
126
USD
AA47142-03
AA47142-06
AA47142-MD
Tin bis(trifluoromethylsulfonyl)imide
Tin bis(trifluoromethylsulfonyl)imide
Tin bis(trifluoromethylsulfonyl)imide
Molecular Formula: C4F12N2SnO8S4
Formula Weight: 678.98
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Off-white
UN No.: 3261
MDL No.: MFCD28100488
Formula:
C₄F₁₂N₂SnO₈S₄ MW: 678.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28100488 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...