Nickelbis(trifluoromethylsulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel bis(trifluoromethanesulphonimide), Nickel bis(trifluoromethanesulphonyl)imide, Nickel bis(trifluoromethylsulphonyl)imide
047102-03
047102-06
047102-MD
AA47102-03EA
133.84
USD
AA47102-03
AA47102-06
AA47102-MD
Nickelbis(trifluoromethylsulfonyl)imide
Nickelbis(trifluoromethylsulfonyl)imide
Nickel bis(trifluoromethylsulfonyl)imide
CAS No.: 207861-63-0
Molecular Formula: C4F12N2NiO8S4
Formula Weight: 618.96
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Green
UN No.: 3261
MDL No.: MFCD23380174
Formula:
C₄F₁₂N₂NiO₈S₄ MW: 618.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23380174 CAS Number: 207861-63-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...