Strontium bis(trifluoromethylsulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Strontium bis(trifluoromethylsulfonyl)imide
CAS No.: 862121-57-1
Molecular Formula: C4F12N2O8S4Sr
Formula Weight: 647.91
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point:
Formula:
C₄F₁₂N₂O₈S₄Sr Melting Pt: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD23380176 CAS Number: 862121-57-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...