AMPIGENE® HS Taq Mix, Enzo Life Sciences
Supplier: Enzo Life Sciences
NUC101-0040 NUC101-0200 NUC101-1000
10014-622EA 63.22 USD
10014-622 10014-624 10014-626
AMPIGENE® HS Taq Mix, Enzo Life Sciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Increased sensitivity standard PCR mix for a broader range of samples, with enhanced speed, yield, and specificity.
For standard PCR applications that require higher sensitivity.
AMPIGENE® HS Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE® HS Taq Mix is a robust mix using hot-start technology to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. An optimized buffer system allows efficient amplification under fast and standard cycling conditions. The AMPIGENE® HS Taq Mix is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine.
Información para pedidos: Contains 2x AMPIGENE® HS Taq Mix (AMPIGENE® HS Taq Polymerase, 6mM MgCl2, 2mM dNTPs, enhancer, and stabilizers).