AMPIGENE® 1-Step RT-PCR Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT105-0050 KIT105-0200 KIT105-1000
10014-584EA 263.78 USD
10014-584 10014-586 10014-588
AMPIGENE® 1-Step RT-PCR Kit, Enzo Life Sciences
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Optimized and efficient cDNA synthesis and PCR in a single tube
For cDNA synthesis and PCR in a single tube.
AMPIGENE® 1-Step RT-PCR Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and PCR in a single tube.The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity.
Información para pedidos: Contains 2x 1-Step Mix (AMPIGENE® HS Taq Polymerase, 6mM MgCl2, 2mM dNTPs, enhancer, and stabilizers), 20x RTase (with RNase inhibitor).