Thermo Scientific™ MAS™ Omni-IMMUNE™ PRO Immunoassay Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
OPRO-SP OPRO-303 OPRO-101 OPRO-202
89209-122PK 283.84 USD
89209-122 89209-120 89209-116 89209-118
Thermo Scientific™ MAS™ Omni-IMMUNE™ PRO Immunoassay Controls, Thermo Scientific
Controls and Calibration Verification Materials
Consolidate immunoassay and tumor marker controls into a single QC product with MAS™ Omni-IMMUNE™ PRO Immunoassay Controls
Liquid, ready-to-use; Human-based serum matrix; Value assignment provided for most major analyzers.
Analytes: 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, ACTH (adrenocorticotropic hormone), Aldosterone, Alpha-fetoprotein, Amikacin, Anti-Tg, Anti-TPO, Benzodiazepine, Beta-2-microglobulin, Beta-HCG, CA 125, CA 15-3, CA 19-9, Carbamazepine, CEA (carcinoembryonic antigen), CKMB, Cortisol, C-Peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol, Free, Ethosuximide, Ferritin, Folate (vitamin B9), Fructosamine, FSH (follicle stimulating hormone), Gastrin, Gentamicin, Growth Hormone, HCG (human chorionic gonadotropin), Homocysteine, IgE, IGF-1 (insulin-like growth factor 1), Inhibin A, Insulin, LH (luteinizing hormone), Lidocaine, Lithium, NAPA (N-acetylprocainamide), PAP (prostatic acid phosphatase), Phenobarbital, Phenytoin, Phenytoin, Free, Primidone, Procainamide, Procalcitonin, Progesterone, Prolactin, PSA Free, PSA Total (prostate-specific antigen), PTH (parathyroid hormone), Quinidine, Salicylate, SHBG (sex hormone-binding globulin), T3 Free, T3 Total (triiodothyroxine), T4 Free, T4 Total (thyroxine), TBG (thyroxine-binding globulin), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, TSH (thyroid-stimulating hormone), T-Uptake, Valproic Acid, Valproic Acid, Free, Vancomycin, Vitamin 12 (refer to package insert for specific analyte and stability claims).
This QC product contains the same analyte panel as the Omni-IMMUNE™ control but also includes the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO), and Sex Hormone Binding Globulin (SHBG).
Información para pedidos: Available in individual levels or Multi-pak.