Thermo Scientific™ MAS™ Omni-IMMUNE™ Immunoassay Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
OIM-101 OIM-202 OIM-303 OIM-SP
89209-108PK 1048.76 USD
89209-108 89209-110 89209-112 89209-114
Thermo Scientific™ MAS™ Omni-IMMUNE™ Immunoassay Controls, Thermo Scientific
Controls and Calibration Verification Materials
Consolidate routine immunoassay, tumor marker, and specialty immunoassay QC processing into a single product with MAS™ Omni-IMMUNE™ Immunoassay Controls.
Liquid, ready-to-use; Human serum based matrix; Value assignment provided for most major analyzer.
Analytes: 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, ACTH (adrenocorticotropic hormone), Aldosterone, Alpha-fetoprotein, Amikacin, Benzodiazepine, Beta-2-microglobulin, Beta-HCG, CA 125, CA 15-3, CA 19-9, Carbamazepine, CEA (carcinoembryonic antigen), CKMB, Cortisol, C-Peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol, Free, Ethosuximide, Ferritin, Folate (vitamin B9), Fructosamine, FSH (follicle stimulating hormone), Gastrin, Gentamicin, Growth Hormone, HCG (human chorionic gonadotropin), Homocysteine, IgE, IGF-1 (insulin-like growth factor 1), Inhibin A, Insulin, LH (luteinizing hormone), Lidocaine, Lithium, NAPA (N-acetylprocainamide), PAP (prostatic acid phosphatase), Phenobarbital, Phenytoin, Phenytoin, Free, Primidone, Procainamide, Procalcitonin, Progesterone, Prolactin, PSA Free, PSA Total (prostate-specific antigen), PTH (parathyroid hormone), Quinidine, Salicylate, T3 Free, T3 Total (triiodothyroxine), T4 Free, T4 Total (thyroxine), TBG (thyroxine-binding globulin), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, TSH (thyroid-stimulating hormone), T-Uptake, Valproic Acid, Valproic Acid, Free, Vancomycin, Vitamin B1 (refer to package insert for specific analyte and stability claims).
Información para pedidos: Available in individual levels or Multi-pak.