Weighing Funnels, Powder, Chemglass
Supplier: Chemglass
CG-1760-10 CG-1760-30 CG-1760-02 CG-1760-06 CG-1760-08 CG-1760-12 CG-1760-18 CG-1760-20 CG-1760-26 CG-1760-28 CG-1760-14 CG-1760-24 CG-1760-16 CG-1760-22
89028-740EA 135.72 USD
89028-740 89028-760 89028-732 89028-736 89028-738 89028-742 89028-748 89028-750 89028-756 89028-758 89028-743 89028-755 89028-747 89028-753
Weighing Funnels, Powder, Chemglass
Boats Weighing Boats
Weighing funnels feature flat bottoms and are designed to fit narrow neck vessels.
They are available in disposable anti-static polypropylene, polypropylene, and borosilicate glass formats.