Atraumatic Occlusion Tools, Edwards
Supplier: Edwards
CPARAL6 CSOFT6 DSAFE61 HCDSOFT8 HYDRA61 SOFT86 SOFT33 CPARAL12
68300-058CS 638.9 USD
68300-058 68300-060 68300-067 68300-074 68300-080 68300-090 68300-086 68300-056
Atraumatic Occlusion Tools, Edwards
Clamps General Purpose Clamps
The Edwards complete line of clamp inserts, spring clips and handleless clamps provide a variety of solutions for atraumatic vessel occlusion.
  • Provides consistent degree of tension
  • Also provide an ideal combination of resilience and traction
The Edwards EverClip reduced-force spring clip minimizes vessel trauma while maintaining occlusion. Clamp inserts are available in 33mm, 61mm and 86mm sizes for the widest range of procedures.
The combinations of atraumatic jaw surfaces offer a range of resilience and traction needed for distinct vessel conditions and procedures.
Handleless clamps incorporate a ratchet-type mechanism that can be adjusted for the precise degree of pressure desired.