JACKSON SAFETY® V60 30-06™ RX Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
19872 19874 19875 20553 20558 20559
10160-692CS 160.4 USD
10160-692 10160-694 10160-696 10160-698 10160-700 10160-702
JACKSON SAFETY® V60 30-06™ RX Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
The JACKSON SAFETY® V60 30-06 RX safety eyewear provide stylish, ergonomic, nylon frame.
  • 'Cheaters' style with diopters for vision assistance
  • Soft, comfortable, self-adjusting, rubber nose pad
  • Brown lenses (safety sunglasses) are ideal for outdoor use
This model comes with brown lenses (safety sunglasses), a tortoise frame and +1.5 Diopters. The lenses are hard coated for longer life and scratch resistance. Made of durable polycarbonate lenses, they provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss.
Both men and women will find these stylish glasses to be comfortable and functional for both vision correction and eye protection.
Base curve 7.
Certificaciones: ANSI Z87.1-2010 Standard.