Digoxigenin NHS ester ≥80% (by HPLC)

Supplier: Enzo Life Sciences

ENZ-45022-0001 ENZ-45022-0005
10014-502EA 271.41 USD
10014-502 10014-504
Digoxigenin NHS ester ≥80% (by HPLC)
Digoxigenin NHS ester
Formula: C₃₅H₅₀N₂O₁₀
Storage Temperature: Freezer
CAS Number: 129273-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da7e806a255287bd5735ea193c64a4847454c08a&vid=30ff9897a62482819e3d5a65c8707e84f5802ec4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR