MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
Supplier: Enzo Life Sciences
950-020-0005 950-020-0025 950-020-0050
10014-536EA 216.91 USD
10014-536 10014-538 10014-540
MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
Slides Microarray Slides
MULTIVIEW® Universal tissue microarray with 11 different human tissue specimens for a broad-spectrum of immunohistochemical staining procedures.