Quartz Crucibles and Lids, Technical Glass Products
Supplier: Technical Glass Products
FQVWR10CRU FQVWR15CRU FQVWR20CRU FQVWR30CRU FQVWR50CRU FQVWR100CRU FQVWR150CRU FQVWR10LID FQVWR15LID FQVWR20LID FQVWR30LID FQVWR50LID FQVWR100LID FQVWR150LID
89049-606EA 89.46 USD
89049-606 89049-608 89049-610 89049-612 89049-614 89049-616 89049-618 89049-620 89049-622 89049-624 89049-626 89049-628 89049-630 89049-632
Quartz Crucibles and Lids, Technical Glass Products
Crucibles
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Quartz crucibles and lids are invaluable in research and industry. Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These properties include; extremely high purity, high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Información para pedidos: All lids are sold separately.