Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 61D3]

Supplier: Tonbo Biosciences
50-0149-T025 50-0149-T100
10753-778EA 82.5 USD
10753-778 10753-780
Anti-CD14 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 61D3]
Antibodies
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR