Qualitative Filter Papers, Advantec MFS
Supplier: Advantec MFS
N0232.0CM N013124.0CM N01319.0CM N0228.5CM N0215.0CM N02600MM N0212.5CM N023532.0CM N0211.0CM N023215.0CM N02319.0CM N023111.0CM N023112.5CM N023115.0CM N014.25CM N015.5CM N017.0CM N019.0CM N0111.0CM N0112.5CM N0115.0CM N0118.5CM N024.25CM N025.5CM N027.0CM N029.0CM N0218.5CM N02314.25CM N02315.5CM N02317.0CM N023118.5CM N02324.25CM N02325.5CM N02327.0CM N02329.0CM N023211.0CM N023212.5CM N023218.5CM N01315.5CM N01317.0CM N013111.0CM N013112.5CM N013115.0CM N013118.5CM N02355.5CM N02357.0CM N02359.0CM N023511.0CM N023512.5CM N023515.0CM N023518.5CM N01015.5CM N01017.0CM N01019.0CM N010111.0CM N010112.5CM N010115.0CM N010118.5CM
101102-326PK 111.29 USD
101102-326 101102-350 101102-414 101102-324 101102-570 101104-918 101102-526 101103-720 101102-444 101105-294 102900-436 102900-438 102900-440 102900-442 102900-444 102900-446 102900-448 102900-450 102900-452 102900-454 102900-456 102900-458 102900-460 102900-462 102900-464 102900-466 102900-468 102900-470 102900-472 102900-474 102900-476 102900-478 102900-480 102900-482 102900-484 102900-486 102900-488 102900-490 102900-492 102900-494 102900-496 102900-498 102900-500 102900-502 102900-504 102900-506 102900-508 102900-510 102900-512 102900-514 102900-516 102900-518 102900-520 102900-522 102900-524 102900-526 102900-528 102900-530
Qualitative Filter Papers, Advantec MFS
Filters Filter Paper Qualitative Filter Paper
Advantec Qualitative Filter Papers are made of 100% cotton linter cellulose.
Grade No. 1 Retains coarse and gelatinous ppt with fast flow rate. Recommended for gas filtration, air pollution analysis, and general purpose. Grade No. 2 Retains medium crystalline ppt with high flow rate. General purpose filter. Grade No. 231 Retains crystalline ppt, moderate flow. Thin and slightly more retentive than No. 1 general purpose paper. Grade No. 232 Collects medium to medium-fine ppt. Slow flow rate. Thin retentive paper. Grade No. 131 High retention efficiency for fine crystalline ppt like BaSO4. Slow flow. Higher wet strength and will withstand suction. Grade No. 235 Highest retention efficiency collecting very fine ppt. Very slow flow rate. Grade No. 101 Creped filter paper captures large particles. Fast flow rate. Excellent for seed germination and viscous fluids .
Primarily used for clarifying and removing precipitates (ppt) and preparation for quantitative analysis. These papers can be used over a pH range from 0-12 and a maximum temperature of 120oC. Ash content of 0.1%. Papers have smooth surface, normal hardness (except No. 101 has creped surface).
Información para pedidos: Other sizes available.