Clarity Hb Check System, Clarity Diagnostics
Supplier: Clarity Diagnostics
CLA-HB4 CLA-HBS100 CLA-HBPROMO1 CLA-HB2
10143-100BX 85.16 USD
10143-100 10143-102 10143-104 10143-622
Clarity Hb Check System, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
For testing hemoglobin and estimated hematocrit in blood
  • Easy to use
  • Large visual display
  • Fast and accurate
  • Results in 15 seconds
  • Small, 10 µL sample size
  • Portable, hand-held, battery-operated device
  • Convenient packaging
  • Ten test cartridges per vial
  • Minimal maintenance
  • Ninety-day open-vial stability of test cartridges
Información para pedidos: The Clarity Hb Check Complete Kit contains one meter, two optical verifiers, logbook, user's manual, quick reference guide and carrying case.