Acetonitrile ≥99.8% for LC-MS
Supplier: Thermo Scientific Chemicals
|
Danger
|
- Liquid
Purity: 99.8%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Appearance: Colorless
Odor: Pungent
Melting Point: -48° to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
UN No.: 1648
MDL No.: MFCD00001878
Applications: Solvent for LC-MS analysis
Formula:
H₃CCN Boiling Pt: 80…82 °C Melting Pt: –48 °C… –45 °C Density: 0.786 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II Merck Index: 14,70 |
|
Specification Test Results
Assay Minimum | 99.8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...