Aluminium ≥99% (metals basis), spheres, Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
042030-KN
042030-KP
AA42030-KMEA
27.19
USD
AA42030-KM
AA42030-KN
AA42030-KP
Aluminium ≥99% (metals basis), spheres, Diameter 6.35 mm (0.25 in)
Aluminium
Purity: 99%
CAS No.: 7429-90-5
Molecular Formula: Alloy 1100, Al:Cu; 99+:0.12 wt%
MDL No.: MFCD00213972
Formula:
Al MW: 26.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...