Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), Thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
039710-GH
039710-FI
AA39710-GHEA
877
USD
AA39710-GH
AA39710-FI
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), Thickness 0.025 mm (0.001 in)
Hafnium
- ~3.3g/100x100mm
Purity: 99.5%
CAS No.: 7440-58-6
Physical Form: ≈3.3g/100x100mm
Odor: Odorless
MDL No.: MFCD00011032
Formula:
Hf MW: 178.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Specification Test Results
Assay Minimum | 99.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...