HEPES-Buffered Saline (5X, pH 7.4), Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J67502-AE J67502-AK
AAJ67502-AEEA 124.64 USD
AAJ67502-AE AAJ67502-AK
HEPES-Buffered Saline (5X, pH 7.4), Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers

Liquid


Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR