1,9-Pyrazoloanthrone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Solid
Purity: 99%
CAS No.: 129-56-6
Molecular Formula: C14H8N2O
Formula Weight: 220.23
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Yellow to brown
Solubility: Soluble in DMSO at approx. 20mg/ml
MDL No.: MFCD00022289
Formula:
C₁₄H₈N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00022289 CAS Number: 129-56-6 EINECS: 204-955-6 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...