ZM-306416 hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

J67265-MA J67265-MCR
AAJ67265-MAEA 310 USD
AAJ67265-MA AAJ67265-MCR
ZM-306416 hydrochloride 98%
ZM-306416 hydrochloride

Solid


Purity: 98% CAS No.: 690206-97-4 Molecular Formula: C16H13N3O2FCl.HCl Formula Weight: 370.21 Physical Form: Solid Appearance: Off-white MDL No.: MFCD08703133

Formula: C₁₆H₁₄Cl₂FN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08703133
CAS Number: 196603-47-1

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR