Prostaglandin F2α tris salt 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dinoprost tromethamine salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol (Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)cyclopentyl)hept-5-enoate
J67245-LB0
J67245-MCR
AAJ67245-LB0EA
280
USD
AAJ67245-LB0
AAJ67245-MA
AAJ67245-MCR
Prostaglandin F2α tris salt 99%
Prostaglandin F2alpha tris salt
Solid
Purity: 99%
CAS No.: 38562-01-5
Molecular Formula: C20H34O5.C4H11NO3
Formula Weight: 475.61
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00077863
Formula:
C₂₄H₄₅NO₈ MW: 475.623 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077863 CAS Number: 38562-01-5 EINECS: 254-002-3 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...