Chloroform Molecular_Biology
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
J67241-AP
J67241-K2
J67241-K4
AAJ67241-APEA
130.45
USD
AAJ67241-AP
AAJ67241-K2
AAJ67241-K4
Chloroform Molecular_Biology
Chloroform
Liquid
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.446
Appearance: Colorless
Odor: Characteristic
Melting Point: -63°
Boiling Point: 61°
Beilstein Reference: 1731042
UN No.: 1888
MDL No.: MFCD00000826
Formula:
CHCl₃ MW: 119.38 g/mol Boiling Pt: 61.7 Melting Pt: -63 Density: 1.492 Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1 III Merck Index: 14,2141 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...