Silver 0.02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 30 nm (Abs. 410 nm)

Supplier: Thermo Scientific Chemicals

J67207-AC J67207-AE
AAJ67207-ACEA 78.4 USD
AAJ67207-AC AAJ67207-AE
Silver 0.02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 30 nm (Abs. 410 nm)
Silver

Liquid


CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Yellow MDL No.: MFCD00003397

Formula: Ag
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR