5-Nitro-2-(3-phenylpropylamino)benzoic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Nitro-2-((3-phenylpropyl)amino)benzoic acid, NPPB

J67152-MA J67152-MF
AAJ67152-MAEA 117 USD
AAJ67152-MA AAJ67152-MF
5-Nitro-2-(3-phenylpropylamino)benzoic acid 98%
5-Nitro-2-(3-phenylpropylamino)benzoic acid

Solid


Purity: 98% CAS No.: 107254-86-4 Molecular Formula: C16H16N2O4 Formula Weight: 300.31 Physical Form: Solid Appearance: Yellow Melting Point: 178-180° MDL No.: MFCD00153851

Formula: C₁₆H₁₆N₂O₄
MW: 300.314 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00153851
CAS Number: 107254-86-4
Merck Index: 14,6725

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR