6-Methoxy-2-naphthylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(6-Methoxynaphthalen-2-yl)acetic acid

J67138-LB0 J67138-MA J67138-MF
AAJ67138-LB0EA 69.5 USD
AAJ67138-LB0 AAJ67138-MA AAJ67138-MF
6-Methoxy-2-naphthylacetic acid 99%
6-Methoxy-2-naphthylacetic acid

Crystalline solid


Purity: 99% CAS No.: 23981-47-7 Molecular Formula: C13H12O3 Formula Weight: 216.24 Physical Form: Crystalline solid Melting Point: 171-173° MDL No.: MFCD00033105

Formula: C₁₃H₁₂O₃
MW: 216.236 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00033105
CAS Number: 23981-47-7
EINECS: 245-967-1

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR