Bicine 99+% for electrophoresis

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

J67087-14 J67087-22
AAJ67087-14EA 56.5 USD
AAJ67087-14 AAJ67087-22
Bicine 99+% for electrophoresis
Bicine

Powder


Purity: 99+% CAS No.: 150-25-4 Molecular Formula: C6H13NO4 Formula Weight: 163.17 Physical Form: Powder Appearance: White Melting Point: ca 191° dec. Beilstein Reference: 1769362 MDL No.: MFCD00004295

Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
Melting Pt: ca 191 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1
Merck Index: 14,1201

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR