DMOG 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-[Methoxy(oxo)acetyl]glycine methyll ester, Dimethyloxaloylglycine, Methyl 2-((2-methoxy-2-oxoethyl)amino)-2-oxoacetate, DMOG

J67077-MA J67077-MC J67077-MF
AAJ67077-MAEA 45.3 USD
AAJ67077-MA AAJ67077-MC AAJ67077-MF
DMOG 98%
DMOG

Solid


Purity: 98% CAS No.: 89464-63-1 Molecular Formula: C6H9NO5 Formula Weight: 175.14 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Off-white MDL No.: MFCD05865098

Formula: C₆H₉NO₅
MW: 175.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05865098
CAS Number: 89464-63-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR