D-myo-Inositol-3-phosphate sodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
Ins(3)P1 sodium salt, 3-IP1 sodium salt
J67072-EXZ
J67072-MCR
AAJ67072-EXDEA
219.99
USD
AAJ67072-EXD
AAJ67072-EXZ
AAJ67072-MCR
D-myo-Inositol-3-phosphate sodium salt
D-myo-Inositol-3-phosphate sodium salt
Lyophilized powder
Purity: 98%
Molecular Formula: C6H12NaO9P
Formula Weight: 282.12
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD05864254
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...