Crocein scarlet 7B ≥70% (dye content)

Supplier: Thermo Scientific Chemicals

J66876-03 J66876-06 J66876-14
AAJ66876-03EA 19.4 USD
AAJ66876-03 AAJ66876-06 AAJ66876-14
Crocein scarlet 7B ≥70% (dye content)
Crocein scarlet 7B

Powder


CAS No.: 6226-76-2 Molecular Formula: C24H18N4Na2O7S2 Formula Weight: 584.53 Physical Form: Powder Appearance: Red Melting Point: 181-188° Solubility: Soluble in water MDL No.: MFCD00056588

Formula: C₂₄H₁₈N₄Na₂O₇S₂
Melting Pt: 181…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00056588
CAS Number: 6226-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR