2-Mercaptoethanol 98+% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydroxyethyl mercaptan, β-Mercaptoethanol
J66742-0B
J66742-30
AAJ66742-0BEA
90.8
USD
AAJ66742-0B
AAJ66742-30
2-Mercaptoethanol 98+% for electrophoresis
2-Mercaptoethanol
Liquid
Purity: 98+%
CAS No.: 60-24-2
Molecular Formula: C2H6OS
Formula Weight: 78.13
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.114
Sensitivity: Air Sensitive; Hygroscopic
Appearance: Colorless
Odor: Unpleasant
Melting Point: -100°
Boiling Point: 156-158°
Beilstein Reference: 773648
UN No.: 2966
MDL No.: MFCD00004890
Formula:
HSCH₂CH₂OH MW: 78.14 g/mol Boiling Pt: 157 Melting Pt: -100 Density: 1.114 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004890 CAS Number: 60-24-2 EINECS: 200-464-6 UN: 2966 ADR: 6.1 II Merck Index: 14,5869 |
|
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...