Bisacrylamide 99+% for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

J66710-14 J66710-22 J66710-30
AAJ66710-14EA 54.01 USD
AAJ66710-14 AAJ66710-22 AAJ66710-30
Bisacrylamide 99+% for electrophoresis
Bisacrylamide

Powder


Purity: 99+% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Physical Form: Powder Density: 1.235 Sensitivity: Air and Light Sensitive Appearance: White Melting Point: 300° Solubility: Soluble in water or methanol Beilstein Reference: 1706297 MDL No.: MFCD00008625

Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Melting Pt: >300 °C
Density: 1.235
Storage Temperature: Ambient
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR