D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
Supplier: Thermo Scientific Chemicals
J66461-EXD
J66461-EXZ
AAJ66461-EXDEA
461
USD
AAJ66461-EXD
AAJ66461-EXZ
D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
Lyophilized powder
Purity: 98%
CAS No.: 210488-61-2
Molecular Formula: C6H8Na8O18P4
Formula Weight: 675.93
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00153818
Formula:
C₆H₈Na₈O₁₈P₄ Storage Temperature: Freezer |
MDL Number:
MFCD00153818 CAS Number: 210488-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...