N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate

J66265-06 J66265-14
AAJ66265-06EA 99 USD
AAJ66265-06 AAJ66265-14
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)

Powder


Applications: For protein solubilization


Applications: For protein solubilization

Formula: C₁₇H₃₇NO₃S
MW: 335.55 g/mol
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036909
CAS Number: 14933-08-5
EINECS: 239-002-3
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR