Gossypol acetic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63517-MC
J63517-MD
AAJ63517-MCEA
77.8
USD
AAJ63517-MC
AAJ63517-MD
Gossypol acetic acid
Gossypol acetic acid
Powder
CAS No.: 12542-36-8
Molecular Formula: C30H30O8.C2H4O2
Formula Weight: 578.61
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow to green
MDL No.: MFCD00058385
Formula:
C₃₂H₃₄O₁₀ Storage Temperature: Refrigerator |
MDL Number:
MFCD00058385 CAS Number: 12542-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...