Ethyl-5-nitroindole-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 5-nitro-1H-indole-2-carboxylate
H66997-03
H66997-06
AAH66997-03EA
42.6
USD
AAH66997-03
AAH66997-06
Ethyl-5-nitroindole-2-carboxylate 97%
Ethyl-5-nitroindole-2-carboxylate
Purity: 97%
CAS No.: 16732-57-3
Molecular Formula: C11H10N2O4
Formula Weight: 234.21
Physical Form: Solid
Beilstein Reference: 225290
MDL No.: MFCD00216477
Formula:
C₁₁H₁₀N₂O₄ MW: 234.211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216477 CAS Number: 16732-57-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...