4-Bromoindole-3-carboxyaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66964-03
H66964-06
AAH66964-03EA
81.2
USD
AAH66964-03
AAH66964-06
4-Bromoindole-3-carboxyaldehyde 97%
4-Bromoindole-3-carboxyaldehyde
Purity: 97%
CAS No.: 98600-34-1
Molecular Formula: C9H6BrNO
Formula Weight: 224.06
Physical Form: Solid
Beilstein Reference: 5733778
MDL No.: MFCD05864695
Formula:
C₉H₆BrNO MW: 224.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864695 CAS Number: 98600-34-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...