Ethyl-3-bromo-1H-indole-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-bromoindole-2-carboxylate, Ethyl 3-Bromoindole-2-carboxylate, 3-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 3-bromo-1H-indole-2-carboxylate

H66951-03 H66951-06
AAH66951-03EA 81.2 USD
AAH66951-03 AAH66951-06
Ethyl-3-bromo-1H-indole-2-carboxylate 97%
Ethyl-3-bromo-1H-indole-2-carboxylate

Purity: 97% CAS No.: 91348-45-7 Molecular Formula: C11H10BrNO2 Formula Weight: 268.11 Physical Form: Solid Beilstein Reference: 186192 MDL No.: MFCD02071793

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02071793
CAS Number: 91348-45-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR