N,N',N''-Trimethyldiethylenetriamine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2'-Bis(methylamino)-N-methyldiethylamine, N,N-Bis[2-(methylamino)ethyl]methylamine, N1,N2-Dimethyl-N1-(2-(methylamino)ethyl)ethane-1,2-diamine
H66905-03
H66905-MD
AAH66905-03EA
340
USD
AAH66905-03
AAH66905-MD
N,N',N''-Trimethyldiethylenetriamine 95%
N,N',N''-Trimethyldiethylenetriamine
Purity: 97%
CAS No.: 105-84-0
Molecular Formula: C7H19N3
Formula Weight: 145.25
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 1736861
MDL No.: MFCD00025637
Formula:
C₇H₁₉N₃ MW: 145.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00025637 CAS Number: 105-84-0 EINECS: 203-337-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...