Methyl-3-amino-4-hydroxybenzoate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-amino-4-hydroxybenzoate

H66894-03 H66894-06
AAH66894-03EA 75.6 USD
AAH66894-03 AAH66894-06
Methyl-3-amino-4-hydroxybenzoate 95%
Methyl-3-amino-4-hydroxybenzoate

Purity: 95% CAS No.: 536-25-4 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Physical Form: Solid Melting Point: 141-143° Beilstein Reference: 5424619 MDL No.: MFCD06659667

Formula: C₈H₉NO₃
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD06659667
CAS Number: 536-25-4
EINECS: 208-627-3

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR