2,5-Dimethylindole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66874-03
H66874-06
AAH66874-03EA
63.8
USD
AAH66874-03
AAH66874-06
2,5-Dimethylindole 97%
Formula:
C₁₀H₁₁N Melting Pt: 112…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005621 CAS Number: 1196-79-8 EINECS: 214-816-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...