6-Bromo-1H-pyrrolo[2,3-b]pyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Bromo-7-azaindole

H66863-03 H66863-06
AAH66863-03EA 186 USD
AAH66863-03 AAH66863-06
6-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
6-Bromo-1H-pyrrolo[2,3-b]pyridine

Purity: 96% CAS No.: 143468-13-7 Molecular Formula: C7H5BrN2 Formula Weight: 197.04 Physical Form: Solid Appearance: Off-white to yellow Melting Point: 192-195° Beilstein Reference: 5424619 UN No.: 2811 MDL No.: MFCD06659667

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 192…195 °C
Storage Temperature: Ambient
MDL Number: MFCD06659667
CAS Number: 143468-13-7
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR