Benzyl viologen dichloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H66836-03 H66836-06
AAH66836-03EA 96.2 USD
AAH66836-03 AAH66836-06
Benzyl viologen dichloride 97%
Benzyl viologen dichloride

Purity: 97% CAS No.: 1102-19-8 Molecular Formula: C24H22Cl2N2 Formula Weight: 409.35 Physical Form: Solid Appearance: White to off-white Melting Point: 260° dec. Beilstein Reference: 3811585 MDL No.: MFCD00011991

Formula: C₂₄H₂₂Cl₂N₂
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011991
CAS Number: 1102-19-8
EINECS: 214-158-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR