Ethyl-5-bromo-1H-indole-2-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 5-bromoindole-2-carboxylate, 5-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 5-Bromoindole-2-carboxylate, Ethyl 5-bromo-1H-indole-2-carboxylate
H66797-03
H66797-06
AAH66797-03EA
34.5
USD
AAH66797-03
AAH66797-06
Ethyl-5-bromo-1H-indole-2-carboxylate 97%
Ethyl-5-bromo-1H-indole-2-carboxylate
Purity: 97%
CAS No.: 16732-70-0
Molecular Formula: C11H10BrNO2
Formula Weight: 268.11
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 163-167°
Beilstein Reference: 170256
MDL No.: MFCD00022701
Formula:
C₁₁H₁₀BrNO₂ MW: 268.11 g/mol Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022701 CAS Number: 16732-70-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...