Methyl indole-7-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 1H-indole-7-carboxylate

H66729-03 H66729-06
AAH66729-03EA 35.2 USD
AAH66729-03 AAH66729-06
Methyl indole-7-carboxylate 97%
Methyl indole-7-carboxylate

Purity: 97% CAS No.: 93247-78-0 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Physical Form: Solid Beilstein Reference: 4402170 MDL No.: MFCD00211064

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211064
CAS Number: 93247-78-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR