5,6-Dihydroxyindole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66709-03
H66709-06
AAH66709-03EA
231
USD
AAH66709-03
AAH66709-06
5,6-Dihydroxyindole 95%
5,6-Dihydroxyindole
Purity: 95%
CAS No.: 3131-52-0
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Physical Form: Solid
Beilstein Reference: 122055
MDL No.: MFCD00798933
Formula:
C₈H₇NO₂ MW: 149.149 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798933 CAS Number: 3131-52-0 EINECS: 412-130-9 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...